Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,4-Trimethylpentane, 95.0%, MilliporeSigma™

Catalog No. 50947639 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-947-639 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-947-639 Supplier MilliporeSigma Supplier No. TX13953
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.23
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specifications

Melting Point −107°C
Quantity 20 L
Flash Point −12°C
Packaging Steel Drum
Assay Percent Range 95.0% min
Vapor Pressure 51 hPa (20°C)
Boiling Point 99°C (1013 hPa)
Solubility Information 0.56 mg/L
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.