Learn More
Selleck Chemical LLC 2.2:5.2-Terthiophene S4982-25mg
SDP
Catalog No. 503108809
Encompass_Preferred
2 2 5 2 -Terthiophene is a natural product occurring in marigold which shows UV-enhanced antibiotic activity
Catalog No. 50-310-8809 Supplier Selleck Chemical LLC Supplier No. S498225MG
2 2 5 2 -Terthiophene is a natural product occurring in marigold which shows UV-enhanced antibiotic activity