Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2':5',2″-Terthiophene, 99%

Catalog No. AAA1674803
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1674803 1 g
AAA16748MD 250 mg
AAA1674806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1674803 Supplier Thermo Scientific Chemicals Supplier No. A1674803
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 1081-34-1 | C12H8S3 | 248.376 g/mol

2,2?:5?,2??-Terthiophene undergoes electrochemical copolymerization along with carbazole and in sodium perchlorate/acetonitrile was reported. Electrochromic copolymer based on TTh and 3, 4-ethylenedioxythiophene has been reported. It acts as a monomer precursor for polythiophene and as a dopant for polycarbonate. It may function as a photosensitizer. It also exihibits antifungal activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′:5′,2′′-Terthiophene undergoes electrochemical copolymerization along with carbazole and in sodium perchlorate/acetonitrile was reported. Electrochromic copolymer based on TTh and 3, 4-ethylenedioxythiophene has been reported. It acts as a monomer precursor for polythiophene and as a dopant for polycarbonate. It may function as a photosensitizer. It also exihibits antifungal activity.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1081-34-1
Molecular Formula C12H8S3
Molecular Weight (g/mol) 248.376
MDL Number MFCD00012167
InChI Key KXSFECAJUBPPFE-UHFFFAOYSA-N
Synonym 2,2':5',2-terthiophene, alpha-terthienyl, alpha-terthiophene, 2,5-di 2-thienyl thiophene, 2,2',5',2-terthienyl, 2,5-bis 2-thienyl thiophene, unii-0p77rau2rr, 2,2'-5',2-terthiophene, 0p77rau2rr, 2,2':5',2-terthienyl
PubChem CID 65067
ChEBI CHEBI:10335
IUPAC Name 2,5-dithiophen-2-ylthiophene
SMILES C1=CSC(=C1)C2=CC=C(S2)C3=CC=CS3

Specifications

Melting Point 89°C to 94°C
Boiling Point 160°C (0.1 mmHg)
Quantity 1 g
Beilstein 178604
Merck Index 14,9174
Solubility Information Insoluble in water.
Formula Weight 248.39
Percent Purity 99%
Chemical Name or Material 2,2':5',2''-Terthiophene
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
RTECSNumber WZ9717750
TSCA No

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.