Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,5,5-Tetramethyl-4-methylene-3-formyl-imidazolidine-1-oxyl, Free Radical 98%, Thermo Scientific™

Catalog No. AC133201000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC133201000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC133201000 Supplier Thermo Scientific Chemicals Supplier No. 133201000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82814-77-5
Molecular Formula C9H16N2O2
Molecular Weight (g/mol) 184.239
InChI Key VLJZUPMHYLYUJK-UHFFFAOYSA-N
Synonym 3-hydroxy-2,2,4,4-tetramethyl-5-methylene-1-imidazolidinecarbaldehyde, 1-imidazolidinyloxy,3-formyl-2,2,5,5-tetramethyl-4-methylene, 3-hydroxy-2,2,4,4-tetramethyl-5-methylene-imidazolidine-1-carbaldehyde
PubChem CID 379377
IUPAC Name 3-hydroxy-2,2,4,4-tetramethyl-5-methylideneimidazolidine-1-carbaldehyde
SMILES CC1(C(=C)N(C(N1O)(C)C)C=O)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 88°C to 93°C
Color Orange
Infrared Spectrum Authentic
Quantity 100 mg
Formula Weight 183.23
Percent Purity ≥97.5%
Physical Form Crystalline Solid
Chemical Name or Material 2,2,5,5-Tetramethyl-4-methylene-3-formyl-imidazolidine-1-oxyl, 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.