Learn More
Medchemexpress LLC HY-111024 100mg Medchemexpress, 2,2,5,7,8-Pentamethyl-6-Chromanol CAS:950-99-2 Purity:>98%
SDP
Catalog No. 501932352
Encompass_Preferred
Medchemexpress, HY-111024 100mg 2,2,5,7,8-Pentamethyl-6-Chromanol CAS:950-99-2 2,2,5,7,8-Pentamethyl-6-Chromanol (PMC) is the anti-oxidant moiety of vitamin E (α-tocopherol). 2,2,5,7,8-Pentamethyl-6-Chromanol has potent androgen receptor (AR) signaling modulation and anti-cancer activity against prostate cancer cell lines [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2352 Supplier Medchemexpress LLC Supplier No. HY111024100MG
Medchemexpress, HY-111024 100mg 2,2,5,7,8-Pentamethyl-6-Chromanol CAS:950-99-2 2,2,5,7,8-Pentamethyl-6-Chromanol (PMC) is the anti-oxidant moiety of vitamin E (α-tocopherol). 2,2,5,7,8-Pentamethyl-6-Chromanol has potent androgen receptor (AR) signaling modulation and anti-cancer activity against prostate cancer cell lines [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.