Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2,5-Dimethyl-1,3-thiazol-4-yl)acetic acid, 97%, Thermo Scientific™

Catalog No. TL01023CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
TL01023CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TL01023CB Supplier Thermo Scientific Chemicals Supplier No. TL01023CB
Add to Cart

Chemical Identifiers

CAS 306937-38-2
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.214
MDL Number MFCD02677725
InChI Key FFPWICPYXBDRHM-UHFFFAOYSA-N
Synonym 2-2,5-dimethylthiazol-4-yl acetic acid, 2-2,5-dimethyl-1,3-thiazol-4-yl acetic acid, 2,5-dimethyl-1,3-thiazol-4-yl acetic acid, 4-thiazoleacetic acid,2,5-dimethyl, dimethyl-1,3-thiazol-4-yl acetic acid, 2,5-dimethyl-thiazol-4-yl-acetic acid, 2-2,5-dimethyl-4-thiazolyl acetic acid, 2-dimethyl-1,3-thiazol-4-yl acetic acid, maybridge3_007546, 4-thiazoleaceticacid,2,5-dimethyl
PubChem CID 2747583
IUPAC Name 2-(2,5-dimethyl-1,3-thiazol-4-yl)acetic acid
SMILES CC1=C(N=C(S1)C)CC(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 171.22
Percent Purity 97%
Chemical Name or Material 2-(2, 5-Dimethyl-1, 3-thiazol-4-yl)acetic acid
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.