Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2,5-Dimethyl-1H-pyrrol-1-yl)benzoic acid, 97%, Thermo Scientific™

Catalog No. CC39901DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC39901DA 1 g
CC39901CB 250 mg
CC39901DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC39901DA Supplier Thermo Scientific Chemicals Supplier No. CC39901DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 92028-57-4
Molecular Formula C13H13NO2
Molecular Weight (g/mol) 215.25
MDL Number MFCD01125264
InChI Key ZLYUUANOICYAAL-UHFFFAOYSA-N
Synonym 2-2,5-dimethyl-1h-pyrrol-1-yl benzoic acid, 2-2,5-dimethylpyrrol-1-yl benzoic acid, 2-2,5-dimethyl-1-pyrrolyl benzoic acid, 2-2,5-dimethyl-pyrrol-1-yl-benzoic acid, 2-2,5-dimethylpyrrolyl benzoic acid, cbmicro_048042, benzoic acid,2-2,5-dimethyl-1h-pyrrol-1-yl
PubChem CID 292956
IUPAC Name 2-(2,5-dimethylpyrrol-1-yl)benzoic acid
SMILES CC1=CC=C(C)N1C1=CC=CC=C1C(O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 215.25
Percent Purity 97%
Chemical Name or Material 2-(2, 5-Dimethyl-1H-pyrrol-1-yl)benzoic acid
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.