Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)acetic acid | 25021-08-3 | MFCD00457254 | 100.0% | 155.11 g·mol⁻¹ | C6H5NO4 | 25 MG
SDP

Catalog No. 5000303955
Encompass_Preferred

2-Maleimido acetic acid is a maleimide-containing carboxylic acid intermediate used in organic synthesis and bioconjugation. It is provided as a high-purity, laboratory-grade solid suitable for research applications.

  • High purity: 99.97% (HPLC).
  • Molecular weight: 155.11 g·mol⁻¹ and formula C6H5NO4.
  • Solubility: soluble in DMSO at 100 mg/mL; ultrasonic agitation recommended.
  • Appearance: white to off-white solid for easy visual identification.
  • Storage: powder stable at -20°C for extended storage; in solution store at -80°C for long term.
  • Analytical confirmation: 1H NMR and LCMS consistent with structure.

Catalog No. 50-003-03955 Supplier Medchemexpress LLC Supplier No. 25MGHYW017831
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Maleimido acetic acid is a maleimide-containing carboxylic acid intermediate used in organic synthesis and bioconjugation. It is provided as a high-purity, laboratory-grade solid suitable for research applications.

  • High purity: 99.97% (HPLC).
  • Molecular weight: 155.11 g·mol⁻¹ and formula C6H5NO4.
  • Solubility: soluble in DMSO at 100 mg/mL; ultrasonic agitation recommended.
  • Appearance: white to off-white solid for easy visual identification.
  • Storage: powder stable at -20°C for extended storage; in solution store at -80°C for long term.
  • Analytical confirmation: 1H NMR and LCMS consistent with structure.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.