Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[2-(5-Methylfuran-2-yl)vinyl]-4,6-bis(trichloromethyl)-1,3,5-triazine 85.0+%, TCI America™
SDP

Catalog No. M12095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M12095G Supplier TCI America Supplier No. M12095G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 156360-76-8
Molecular Formula C12H7Cl6N3O
Molecular Weight (g/mol) 421.908
MDL Number MFCD00671558
InChI Key XOPKKHCDIAYUSK-UHFFFAOYSA-N
Synonym 2,4-Bis(trichloromethyl)-6-[2-(5-methylfuran-2-yl)vinyl]-1,3,5-triazine
PubChem CID 53405094
IUPAC Name 2-[2-(5-methylfuran-2-yl)ethenyl]-4,6-bis(trichloromethyl)-1,3,5-triazine
SMILES CC1=CC=C(O1)C=CC2=NC(=NC(=N2)C(Cl)(Cl)Cl)C(Cl)(Cl)Cl

Specifications

Melting Point 156°C
Quantity 5 g
Formula Weight 421.91
Percent Purity ≥85.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-[2-(5-Methylfuran-2-yl)vinyl]-4,6-bis(trichloromethyl)-1,3,5-triazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.