Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,6,6-Tetrachlorocyclohexanol 98%, Thermo Scientific™

Catalog No. AC181680500
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC181680500 50 g
AC181680100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC181680500 Supplier Thermo Scientific Chemicals Supplier No. 181680500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56207-45-5
Molecular Formula C6H8Cl4O
Molecular Weight (g/mol) 237.93
MDL Number MFCD00003823
InChI Key RORBTKDJFQCFMD-UHFFFAOYSA-N
Synonym 2,2,6,6-tetrachlorocyclohexanol, 2,2,6,6,-tetrachlorocyclohexanol, cyclohexanol, 2,2,6,6-tetrachloro, cyclohexanol,2,2,6,6-tetrachloro
PubChem CID 92061
IUPAC Name 2,2,6,6-tetrachlorocyclohexan-1-ol
SMILES OC1C(Cl)(Cl)CCCC1(Cl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 55°C to 60°C
Color Beige or White
Infrared Spectrum Authentic
Linear Formula Cl4C6H7OH
Quantity 50 g
Formula Weight 237.94
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 2,2,6,6-Tetrachlorocyclohexanol, 98%
Show More Show Less
EINECSNumber 260-055-3
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.