Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2,2,6,6-Tetramethyl-3,5-heptanedionato)lithium, 98%, Thermo Scientific Chemicals

Catalog No. AA3859606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3859606 5 g
AA3859603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3859606 Supplier Thermo Scientific Chemicals Supplier No. 03859606
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 22441-13-0

Used as reactant for synthesis of Bis-cycloiridiated complexes for electroluminescent studies, cationic imido alkylidene complexes, polyaromatic ligands containing pyrene and mono-cyclometalated platinum complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22441-13-0
MDL Number MFCD00142754
Synonym Lithium 2,2,6,6-tetramethyl-3,5-heptanedionate; Li(TMHD)

Specifications

Melting Point 127°C
Boiling Point 180°C (0.05 mmHg)
Quantity 5 g
Solubility Information Soluble in acetone and methanol.
Formula Weight 190.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (2,2,6,6-Tetramethyl-3,5-heptanedionato)lithium
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.