Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,6,6-Tetramethyl-3,5-heptanedione, 98%

Catalog No. AC168321000
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC168321000 100 g Glass bottle
AC168320050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC168321000 Supplier Thermo Scientific Chemicals Supplier No. 168321000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1118-71-4 | C11H20O2 | 184.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1118-71-4
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.28
InChI Key YRAJNWYBUCUFBD-UHFFFAOYSA-N
Synonym dipivaloylmethane, 2,2,6,6-tetramethyl-3,5-heptanedione, 3,5-heptanedione, 2,2,6,6-tetramethyl, unii-r8ui909hoy, tmhd, 2,2,6,6-tetramethyl-3,5-heptanedione dipivaloylmethane, r8ui909hoy, 2,2,6,6-tetramethyl-heptane-3,5-dione, pubchem12497
PubChem CID 70700
IUPAC Name 2,2,6,6-tetramethylheptane-3,5-dione
SMILES CC(C)(C)C(=O)CC(=O)C(C)(C)C

Specifications

Color Colorless to Yellow
Density 0.8800g/mL
Boiling Point 72.0°C to 73.0°C (6.0 mmHg)
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4579 to 1.4599
Linear Formula (CH3)3CCOCH2COC(CH3)3
Quantity 100 g
Beilstein 01,III,3149
Specific Gravity 0.88
Solubility Information Solubility in water: insoluble.
Formula Weight 184.28
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2,2,6,6-Tetramethyl-3,5-heptanedione
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 214-268-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.