Learn More
Medchemexpress LLC 2,2,6,6-Tetramethyl-4-piperidone | 826-36-8 | 155.24 | 100 G
Supplier: Medchemexpress LLC HYN1131100G
Triacetonamine is an intermediate used in the synthesis of pharmaceutical products, pesticides, and photostabilizers for polymers. It has oral activity and can induce acute liver failure (ALF) in rats.
- Intermediate for synthesizing pharmaceutical products
- Used in pesticide synthesis
- Component in photostabilizers for polymers
- Exhibits oral activity
- Can induce acute liver failure (ALF) in rats