Learn More
Strem, An Ascensus Company CAS# 1118-71-4. 5g. 2,2,6,6-Tetramethylheptane-3,5-dione, 98% TMHD. MFCD00008848
SDP
Catalog No. 5090109267
Encompass
CAS# 1118-71-4. 5g. 2,2,6,6-Tetramethylheptane-3,5-dione, 98% TMHD. MFCD00008848. Molecular Weight: 184.28. Molecular Formula: (CH3)3CC(O)CH2C(O)C(CH3)3. Color/form: colorless to pale yellow liq. or low melting solid Strem# 08-2050. http://www.strem.com/catalog/v/08-2050/
Catalog No. 50-901-09267 Supplier Strem, An Ascensus Company Supplier No. 0820505G
CAS# 1118-71-4. 5g. 2,2,6,6-Tetramethylheptane-3,5-dione, 98% TMHD. MFCD00008848. Molecular Weight: 184.28. Molecular Formula: (CH3)3CC(O)CH2C(O)C(CH3)3. Color/form: colorless to pale yellow liq. or low melting solid Strem# 08-2050. http://www.strem.com/catalog/v/08-2050/