Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,6,6-Tetramethylpimelic Acid 98.0+%, TCI America™
SDP

Catalog No. T18185G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T18185G Supplier TCI America Supplier No. T18185G

Chemical Identifiers

CAS 2941-45-9
Molecular Formula C11H18O4
Molecular Weight (g/mol) 214.26
MDL Number MFCD00800574
InChI Key CKUJONFLPBQEKM-UHFFFAOYSA-L
Synonym 2,2,6,6-tetramethylpimelic acid, acmc-209h8n, heptanedioic acid,2,2,6,6-tetramethyl
PubChem CID 2733156
IUPAC Name 2,2,6,6-tetramethylheptanedioate
SMILES CC(C)(CCCC(C)(C)C([O-])=O)C([O-])=O

Specifications

Melting Point 171°C
Color White
Quantity 5 g
Formula Weight 216.28
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2,6,6-Tetramethylpimelic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.