Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2,6,6-Tetramethylpiperidine

Catalog No. 11575425G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-575-425G 25 g
11-575-45G 5 g
11-575-410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 11-575-425G Supplier Sigma Aldrich Supplier No. 11575425G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The lithium amide has been used to efficiently ortho deprotonate pyridine-3-carboxamides.

Specifications

CAS 768-66-1
Density 0.837 g/mL at 25°C
Boiling Point 152°C
Molecular Formula C9H19N
Linear Formula C9H19N
Refractive Index n20/D 1.445
MDL Number MFCD00005985
Quantity 25 g
Molecular Weight (g/mol) 141.25 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.