Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2,6,6-Tetramethylpiperidinylmagnesium Chloride Lithium Chloride Complex Solution

Catalog No. 703540500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-354-0500ML 500 mL
70-354-0100ML 100 mL
70-354-0 4 x 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 70-354-0500ML Supplier Sigma Aldrich Supplier No. 703540500ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Selective Deprotonations using Knochel-Hauser-BaseSure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 898838-07-8
Density 0.96 g/mL at 25°C
Molecular Formula C9H18Cl2LiMgN
Linear Formula C9H18Cl2LiMgN
MDL Number MFCD12546030
Quantity 500 mL
Synonym TMPMgCl-+LiCl
Molecular Weight (g/mol) 242.4 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.