Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(2,6,7,8-Tetrahydro-1H-indeno[5,4-b]furan-8-yl)ethanamine hydrochloride | 1053239-39-6 | 99.9% | C13H18ClNO | 250 MG
SDP

Catalog No. 5000358438
Encompass_Preferred

This product is 2-(2,6,7,8-Tetrahydro-1H-indeno[5,4-b]furan-8-yl)ethanamine hydrochloride, a drug intermediate used for the synthesis of various active compounds.

  • Appearance: Solid
  • Melting point: 165-167 °C
  • Harmful if swallowed.
  • Causes skin and serious eye irritation.
  • May cause respiratory irritation.
  • Avoid breathing dust/fume/vapours/spray.
  • Wear protective gloves, clothing, and eye/face protection.

Catalog No. 50-003-58438 Supplier Medchemexpress LLC Supplier No. 250MGHY75253A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is 2-(2,6,7,8-Tetrahydro-1H-indeno[5,4-b]furan-8-yl)ethanamine hydrochloride, a drug intermediate used for the synthesis of various active compounds.

  • Appearance: Solid
  • Melting point: 165-167 °C
  • Harmful if swallowed.
  • Causes skin and serious eye irritation.
  • May cause respiratory irritation.
  • Avoid breathing dust/fume/vapours/spray.
  • Wear protective gloves, clothing, and eye/face protection.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.