Learn More
Medchemexpress LLC 2-(2,6,7,8-Tetrahydro-1H-indeno[5,4-b]furan-8-yl)ethanamine hydrochloride | 1053239-39-6 | C13H18ClNO | 5 G
SDP
Catalog No. 5000434222
Encompass_Preferred
2-(2,6,7,8-Tetrahydro-1H-indeno[5,4-b]furan-8-yl)ethanamine hydrochloride is a drug intermediate used for the synthesis of various active compounds. This product appears as a solid with a molecular weight of 239.75 and a melting point of 165-167 °C. It is stable under recommended storage conditions.
  • Drug intermediate for active compound synthesis
  • Recommended storage at 4°C, sealed, away from moisture
  • For solvent solutions, store at -80°C for 6 months or -20°C for 1 month
  • For research use only
Catalog No. 50-004-34222 Supplier Medchemexpress LLC Supplier No. HY75253A5G
2-(2,6,7,8-Tetrahydro-1H-indeno[5,4-b]furan-8-yl)ethanamine hydrochloride is a drug intermediate used for the synthesis of various active compounds. This product appears as a solid with a molecular weight of 239.75 and a melting point of 165-167 °C. It is stable under recommended storage conditions.
  • Drug intermediate for active compound synthesis
  • Recommended storage at 4°C, sealed, away from moisture
  • For solvent solutions, store at -80°C for 6 months or -20°C for 1 month
  • For research use only