Learn More
Medchemexpress LLC 2- 2- 6-chlorohexylo 250mg
SDP
Catalog No. 5000204647
Encompass_Preferred
2-(2-(6-chlorohexyloxy)ethoxy)ethanamine (hydrochloride) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-04647 Supplier Medchemexpress LLC Supplier No. 250MGHYW096093
2-(2-(6-chlorohexyloxy)ethoxy)ethanamine (hydrochloride) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]