Learn More
Medchemexpress LLC 2-[[2,6-dichloro-4-(3,5-dimethyl-4-isoxazolyl)phenyl]amino]-benzoic acid | 2243736-35-6 | MFCD32669807 | >98.0% | C18H14Cl2N2O3 | 10MG
SDP
Catalog No. 5000261301
Encompass_Preferred
FB23 is a potent, selective small-molecule inhibitor of the fat mass and obesity-associated (FTO) mRNA N6-methyladenosine (m6A) demethylase. It binds directly to FTO and inhibits its demethylase activity (IC50 ~60 nM), and has been used in cellular studies to probe FTO function and effects on leukemia cell proliferation.
  • Potent FTO inhibition (IC50 ~60 nM).
  • Selective for FTO over related demethylases.
  • Direct binding to the target demonstrated.
  • Suitable for cellular assays probing m6A regulation and proliferation effects.
  • Supplied as a solid with high purity (>98%).
Catalog No. 50-002-61301 Supplier Medchemexpress LLC Supplier No. HY13718710MG
FB23 is a potent, selective small-molecule inhibitor of the fat mass and obesity-associated (FTO) mRNA N6-methyladenosine (m6A) demethylase. It binds directly to FTO and inhibits its demethylase activity (IC50 ~60 nM), and has been used in cellular studies to probe FTO function and effects on leukemia cell proliferation.
  • Potent FTO inhibition (IC50 ~60 nM).
  • Selective for FTO over related demethylases.
  • Direct binding to the target demonstrated.
  • Suitable for cellular assays probing m6A regulation and proliferation effects.
  • Supplied as a solid with high purity (>98%).