Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

{2-[(2,6-Dichlorophenyl)amino]phenyl}Methanol Pharmaceutical Impurity Standard, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013790 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3790 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3790 Supplier MilliporeSigma Supelco Supplier No. 0471850MG
Add to Cart
Add to Cart

Specifications

CAS 27204-57-5
Linear Formula C13H11 Cl2 NO
MDL Number MFCD03093753
Synonym Diclofenac impurity C (PhEur); 2-[(2,6-Dichlorophenyl)amino]benzenemethanol; o-(2,6-Dichloroanilino)benzyl alcohol
Type Pharmaceutical Impurity Standard
Quantity 50 mg
Formula Weight 268.14
Percent Purity ≥98.0% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.