Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-[(2,6-dichlorophenyl)methylsulfanyl]-5-(1H-pyrrol-2-yl)-1,3,4-oxadiazole | 2253744-54-4 | MFCD32174346 | 99.9% | 326.20 g·mol⁻¹ | C13H9Cl2N3OS | 100MG
SDP

Supplier:  Medchemexpress LLC HY126010100MG

Encompass_Preferred

Dooku1 is a reversible small-molecule antagonist of Yoda1-activated Piezo1 channels used in vascular physiology and ion channel research. It inhibits Yoda1-induced Ca2+ entry with IC50 values of 1.3 μM in HEK293 cells and 1.5 μM in HUVECs, and is supplied as a high-purity solid with solution formats and supporting documentation.

  • Contains CAS number 2253744-54-4.
  • Molecular formula C13H9Cl2N3OS.
  • Molecular weight 326.20 g·mol⁻¹.
  • High purity for research use.
  • Available as solid and as DMSO solution formats.
  • Documentation includes datasheet, COA, and SDS.

Catalog No. 50-002-36384


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.