Learn More
Medchemexpress LLC 2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindoline-4-carboxylic acid | 1547163-38-1 | MFCD32063448 | 98.0% | 302.24 g·mol⁻¹ | C14H10N2O6 | 1 G
SDP
Catalog No. 5000282492
Encompass_Preferred
Thalidomide acid is a carboxylic-acid-functionalized thalidomide analog used as an E3 ligase ligand in targeted protein degradation (PROTAC) research. Supplied as a powder for research use only, it serves as a building block for conjugation and linker attachment in chemical biology and medicinal chemistry workflows.
  • Useful as an E3 ligase ligand for PROTAC development.
  • Carboxylic acid functional group suitable for conjugation chemistry.
  • High purity suitable for research applications.
  • Powder form for ease of handling and storage.
  • Characterized by CAS 1547163-38-1 and molecular weight 302.24 g·mol⁻¹.
Catalog No. 50-002-82492 Supplier Medchemexpress LLC Supplier No. HYW2416351G
Thalidomide acid is a carboxylic-acid-functionalized thalidomide analog used as an E3 ligase ligand in targeted protein degradation (PROTAC) research. Supplied as a powder for research use only, it serves as a building block for conjugation and linker attachment in chemical biology and medicinal chemistry workflows.
  • Useful as an E3 ligase ligand for PROTAC development.
  • Carboxylic acid functional group suitable for conjugation chemistry.
  • High purity suitable for research applications.
  • Powder form for ease of handling and storage.
  • Characterized by CAS 1547163-38-1 and molecular weight 302.24 g·mol⁻¹.