Learn More
Medchemexpress LLC 2-(2,6-dioxopiperidin-3-yl)-4,5-difluoroisoindoline-1,3-dione | 2222115-19-5 | >98.0% | 294.21 g/mol | C13H8F2N2O4 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW5768681G
Encompass_Preferred
Thalidomide-4,5-F is a fluorinated analogue of thalidomide used as a cereblon (CRBN) E3 ligase ligand in research. It promotes ubiquitinylation of target proteins and can be derivatized via substitution at the fluorine positions. Supplied as a powder with high reported purity for use in chemical biology and medicinal chemistry studies.
  • Fluorinated thalidomide analogue enabling derivatization via fluorine substitution
  • Acts as a cereblon (CRBN) E3 ligase ligand for targeted protein ubiquitination
  • Promotes ubiquitinylation of proteins in biochemical assays
  • High reported purity (≥98.0%) suitable for research use
  • Powder form compatible with synthetic and analytical workflows
  • Molecular formula C13H8F2N2O4 and molecular weight 294.21 g/mol
Catalog No. 50-002-84427