Learn More
Medchemexpress LLC 2-(2,6-dioxopiperidin-3-yl)-4,5-difluoroisoindoline-1,3-dione | 2222115-19-5 | >98.0% | 294.21 g·mol⁻¹ | C13H8F2N2O4 | 50 MG

Supplier: Medchemexpress LLC HYW57686850MG
Thalidomide-4,5-F is a fluorinated analogue of thalidomide that serves as a cereblon (CRBN) ligand for E3 ligase recruitment and targeted protein degradation research. It promotes ubiquitinylation of proteins and is suitable for derivatization at its fluorine positions, making it a useful building block in PROTAC and molecular glue chemistries.
- Promotes ubiquitinylation of proteins.
- Useful as a cereblon ligand for E3 ligase recruitment and PROTAC synthesis.
- Fluorine positions amenable to further derivatization.
- High purity (>98.0%).
- Chemical formula C13H8F2N2O4; molecular weight 294.21 g·mol⁻¹.
- Appearance: white to off-white solid; available in multiple sizes.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.