Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(2,6-dioxopiperidin-3-yl)-4-iodoisoindoline-1,3-dione | 959150-64-2 | MFCD31735040 | 99.9% | 384.13 g/mol | C13H9IN2O4 | 250 MG
SDP

Catalog No. 5000208921
Encompass_Preferred

2-(2,6-dioxopiperidin-3-yl)-4-iodoisoindoline-1,3-dione is a thalidomide derivative bearing an iodine substituent on the isoindoline ring, supplied as a purple to brown solid for research use. It is characterized by its molecular formula (C13H9IN2O4), molecular weight, and high HPLC purity, and requires light protection during cold storage.

  • Thalidomide derivative with iodine substituent.
  • High purity (HPLC) suitable for research applications.
  • Purple to brown solid, confirmed by analytical data.
  • Molecular formula C13H9IN2O4 and molecular weight 384.13 g/mol.
  • Store at 4°C and protect from light; solvent stability: -80°C (6 months), -20°C (1 month).
  • Analytical characterization includes HPLC and 1H NMR consistent with structure.

Catalog No. 50-002-08921 Supplier Medchemexpress LLC Supplier No. 250MGHYW076313
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(2,6-dioxopiperidin-3-yl)-4-iodoisoindoline-1,3-dione is a thalidomide derivative bearing an iodine substituent on the isoindoline ring, supplied as a purple to brown solid for research use. It is characterized by its molecular formula (C13H9IN2O4), molecular weight, and high HPLC purity, and requires light protection during cold storage.

  • Thalidomide derivative with iodine substituent.
  • High purity (HPLC) suitable for research applications.
  • Purple to brown solid, confirmed by analytical data.
  • Molecular formula C13H9IN2O4 and molecular weight 384.13 g/mol.
  • Store at 4°C and protect from light; solvent stability: -80°C (6 months), -20°C (1 month).
  • Analytical characterization includes HPLC and 1H NMR consistent with structure.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.