Learn More
Medchemexpress LLC 2-(2,6-dioxopiperidin-3-yl)-5-methylisoindoline-1,3-dione | 40313-92-6 | MFCD30290642 | 99.9% | C14H12N2O4 | 10MG
SDP
Catalog No. 5000260617
Encompass_Preferred
Thalidomide-5-methyl is a thalidomide-derived ligand that recruits the cereblon (CRBN) E3 ubiquitin ligase and is used as a recruiting moiety in proteolysis-targeting chimera (PROTAC) synthesis and CRBN-focused chemical biology research.
  • Binds cereblon (CRBN) E3 ubiquitin ligase.
  • Suitable as a recruiting moiety in PROTAC synthesis.
  • High purity (99.9%).
  • Molecular formula C14H12N2O4, molecular weight 272.26.
  • Provided as a small-molecule solid, common pack size 10 mg.
  • Compatible with linker conjugation for medicinal chemistry applications.
Catalog No. 50-002-60617 Supplier Medchemexpress LLC Supplier No. HY13954810MG
Thalidomide-5-methyl is a thalidomide-derived ligand that recruits the cereblon (CRBN) E3 ubiquitin ligase and is used as a recruiting moiety in proteolysis-targeting chimera (PROTAC) synthesis and CRBN-focused chemical biology research.
  • Binds cereblon (CRBN) E3 ubiquitin ligase.
  • Suitable as a recruiting moiety in PROTAC synthesis.
  • High purity (99.9%).
  • Molecular formula C14H12N2O4, molecular weight 272.26.
  • Provided as a small-molecule solid, common pack size 10 mg.
  • Compatible with linker conjugation for medicinal chemistry applications.