Learn More
Medchemexpress LLC 2-(2,6-Dioxopiperidin-3-yl)phthalimidine | 26581-81-7 | 99.7% | 244.25 | 200 MG
SDP
Catalog No. 5000350497
Encompass_Preferred
2-(2,6-Dioxopiperidin-3-yl)phthalimidine (EM-12) is a teratogenic Thalidomide analogue that is more active than Thalidomide and is more stable for hydrolysis. It enhances 1,2-dimethylhydrazine-induction of rat colon adenocarcinomas. It is an off-white to gray solid.
  • Teratogenic thalidomide analogue
  • More active than thalidomide
  • More stable to hydrolysis
  • Enhances 1,2-dimethylhydrazine-induction of rat colon adenocarcinomas
  • Off-white to gray solid
Catalog No. 50-003-50497 Supplier Medchemexpress LLC Supplier No. HY138793200MG
2-(2,6-Dioxopiperidin-3-yl)phthalimidine (EM-12) is a teratogenic Thalidomide analogue that is more active than Thalidomide and is more stable for hydrolysis. It enhances 1,2-dimethylhydrazine-induction of rat colon adenocarcinomas. It is an off-white to gray solid.
  • Teratogenic thalidomide analogue
  • More active than thalidomide
  • More stable to hydrolysis
  • Enhances 1,2-dimethylhydrazine-induction of rat colon adenocarcinomas
  • Off-white to gray solid