Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,8,8-Tetramethyl-3,6-nonadiyn-5-ol 98.0+%, TCI America™
SDP

Catalog No. T20315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20315G 5 g
T20311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T20315G Supplier TCI America Supplier No. T20315G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50428-39-2
Molecular Formula C13H20O
Molecular Weight (g/mol) 192.302
MDL Number MFCD06797185
InChI Key KGZASKNUKKURJQ-UHFFFAOYSA-N
Synonym 1,5-Di-tert-butyl-1,4-pentadiyn-3-ol, 1,3-Dineopentylidyne-2-propanol
PubChem CID 11355997
IUPAC Name 2,2,8,8-tetramethylnona-3,6-diyn-5-ol
SMILES CC(C)(C)C#CC(C#CC(C)(C)C)O

Specifications

Melting Point 80°C
Color White
Quantity 5 g
Formula Weight 192.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2,8,8-Tetramethyl-3,6-nonadiyn-5-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.