Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(2-(allyloxy)ethoxy)ethanol | 15075-50-0 | MFCD00078367 | 99.3% | 146.18 g/mol | C7H14O3 | 100 MG
SDP

Catalog No. 5000210892
Encompass_Preferred

Allyl-PEG2-OH is a short polyethylene glycol (PEG2) derivative bearing an allyl functional group used as a building block in polymer chemistry, bioconjugation, and drug-delivery research.

  • Used as a linker or reactive handle for polymerization and conjugation
  • Molecular formula: C7H14O3
  • Molecular weight: 146.18 g/mol
  • Purity: 99.33% (GC)
  • Appearance: colorless to light yellow liquid
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)

Catalog No. 50-002-10892 Supplier Medchemexpress LLC Supplier No. HYW047691100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Allyl-PEG2-OH is a short polyethylene glycol (PEG2) derivative bearing an allyl functional group used as a building block in polymer chemistry, bioconjugation, and drug-delivery research.

  • Used as a linker or reactive handle for polymerization and conjugation
  • Molecular formula: C7H14O3
  • Molecular weight: 146.18 g/mol
  • Purity: 99.33% (GC)
  • Appearance: colorless to light yellow liquid
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.