Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Aminoethyl)pyridine, 95%

Catalog No. AC103690010
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103690010 1 g Glass bottle
AC103690100 10 g Glass bottle
AC103690500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC103690010 Supplier Thermo Scientific Chemicals Supplier No. 103690010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2706-56-1 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2706-56-1
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00006367
InChI Key XPQIPUZPSLAZDV-UHFFFAOYSA-N
Synonym 2-2-aminoethyl pyridine, 2-pyridylethylamine, 2-2-pyridyl ethylamine, 2-pyridin-2-yl ethanamine, 2-pyridineethanamine, demethylbetahistine, 2-pyridin-2-yl ethan-1-amine, alpha-pyridylethylamine, pyridine, 2-2-aminoethyl, 2-aminoethylpyridine
PubChem CID 75919
ChEBI CHEBI:74024
IUPAC Name 2-pyridin-2-ylethanamine
SMILES NCCC1=CC=CC=N1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0210g/mL
Boiling Point 105°C to 106°C (10.0 mmHg)
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5340 to 1.538
Linear Formula (C5H4N)CH2CH2NH2
Quantity 1 g
Beilstein 22, 434
Specific Gravity 1.021
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most common polar solvents,insoluble in non-polar solvents and oils
Formula Weight 122.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-(2-Aminoethyl)pyridine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
EINECSNumber 220-295-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.