Learn More
Medchemexpress LLC 2,2'-Anhydrouridine (2,2'-cyclouridine) | 3736-77-4 | 99.5% | 226.19 | 100 G
SDP
Catalog No. 5000452335
Encompass_Preferred
2,2'-Anhydrouridine (2,2'-Cyclouridine) is a nucleoside analog and an important intermediate in the synthesis of other nucleoside-related compounds. The 5-substituted analogues of 2,2'-Anhydrouridine are competitive inhibitors of uridine phosphorylase. This product appears as a solid, white to off-white in color.
  • Nucleoside analog
  • Intermediate in synthesis of nucleoside-related compounds
  • Competitive inhibitor of uridine phosphorylase (5-substituted analogues)
  • Solid appearance
  • White to off-white color
Catalog No. 50-004-52335 Supplier Medchemexpress LLC Supplier No. HYW012313100G
2,2'-Anhydrouridine (2,2'-Cyclouridine) is a nucleoside analog and an important intermediate in the synthesis of other nucleoside-related compounds. The 5-substituted analogues of 2,2'-Anhydrouridine are competitive inhibitors of uridine phosphorylase. This product appears as a solid, white to off-white in color.
  • Nucleoside analog
  • Intermediate in synthesis of nucleoside-related compounds
  • Competitive inhibitor of uridine phosphorylase (5-substituted analogues)
  • Solid appearance
  • White to off-white color