Learn More
Medchemexpress LLC 2-(2-azidoethoxy)ethyl 4-methylbenzenesulfonate | 182347-24-6 | MFCD27946436 | 99.8% | 285.32 g/mol | C11H15N3O4S | 500 MG
SDP
Catalog No. 5000281800
Encompass_Preferred
N3-PEG2-Tos is an azide-terminated PEG2 tosylate linker used as a synthetic building block for organic synthesis and bioconjugation. The tosylate is a good leaving group for nucleophilic substitution, and the azide serves as a click-chemistry handle for rapid, selective conjugation under copper-catalyzed conditions.
  • Azide functional group for copper-catalyzed azide-alkyne cycloaddition.
  • Tosylate leaving group enables efficient nucleophilic substitution.
  • Short PEG2 spacer provides flexibility and improved solubility in common organic solvents.
  • High reported purity supports consistent reaction performance.
  • Available in multiple package sizes to suit different synthesis scales.
Catalog No. 50-002-81800 Supplier Medchemexpress LLC Supplier No. HYW190836500MG
N3-PEG2-Tos is an azide-terminated PEG2 tosylate linker used as a synthetic building block for organic synthesis and bioconjugation. The tosylate is a good leaving group for nucleophilic substitution, and the azide serves as a click-chemistry handle for rapid, selective conjugation under copper-catalyzed conditions.
  • Azide functional group for copper-catalyzed azide-alkyne cycloaddition.
  • Tosylate leaving group enables efficient nucleophilic substitution.
  • Short PEG2 spacer provides flexibility and improved solubility in common organic solvents.
  • High reported purity supports consistent reaction performance.
  • Available in multiple package sizes to suit different synthesis scales.