Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2'-Azobis(2-Methylpropionitrile) Solution

Catalog No. 714887100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-488-7100ML 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 71-488-7100ML Supplier Sigma Aldrich Supplier No. 714887100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,2'sa-Azobis(2-methylpropionitrile) (AIBN) is a radical initiator. AIBN solution can be used to initiate radical-induced reactions, specifically free-radical polymerizations. It can be used in:Synthesis of styrene-vinyl pyridine diblock copolymers by reversible addition-fragmentation chain transfer (RAFT) polymerization.Preparation of silicon oxycarbide glasses.Synthesis of poly [N-(p-vinyl benzyl) phthalimide] for the preparation of titanium dioxide composites for electrophoretic displays.

Specifications

CAS 894085-97-3
Melting Point 127°C to 131°C
Molecular Formula C21H27N2P
Refractive Index n20/D 1.495
Linear Formula C21H27N2P
MDL Number MFCD09038825
Quantity 100 mg
Synonym alpha,alpha,'-Azoisobutyronitrile solution; AIBN solution
Molecular Weight (g/mol) 338.43 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.