Learn More
Medchemexpress LLC 2-(2-benzofuranyl)-2-imidazoline hydrochloride | 89196-95-2 | MFCD00672666 | >=98.0% | 222.67 g/mol | C11H11ClN2O | 1 ML
SDP
Catalog No. 5000212255
Encompass_Preferred
RX 801077 hydrochloride (2-(2-benzofuranyl)-2-imidazoline hydrochloride) is a selective imidazoline I2 receptor (I2R) agonist supplied for research use. It is available as a solid and as a ready-to-use 10 mM solution in DMSO (1 mL), and is commonly used in in vitro pharmacology and receptor-binding studies focused on inflammation and neuroprotection. Consult the safety data sheet for handling and storage.
  • Selective imidazoline I2 receptor agonist.
  • Available as solid powder and as a 10 mM solution in DMSO, 1 mL.
  • High purity suitable for analytical and pharmacological studies.
  • Molecular weight 222.67 g/mol; formula C11H11ClN2O.
  • Intended for in vitro pharmacology, receptor binding, and neuroprotection research.
Catalog No. 50-002-12255 Supplier Medchemexpress LLC Supplier No. HY10090410MM 1ML
RX 801077 hydrochloride (2-(2-benzofuranyl)-2-imidazoline hydrochloride) is a selective imidazoline I2 receptor (I2R) agonist supplied for research use. It is available as a solid and as a ready-to-use 10 mM solution in DMSO (1 mL), and is commonly used in in vitro pharmacology and receptor-binding studies focused on inflammation and neuroprotection. Consult the safety data sheet for handling and storage.
  • Selective imidazoline I2 receptor agonist.
  • Available as solid powder and as a 10 mM solution in DMSO, 1 mL.
  • High purity suitable for analytical and pharmacological studies.
  • Molecular weight 222.67 g/mol; formula C11H11ClN2O.
  • Intended for in vitro pharmacology, receptor binding, and neuroprotection research.