Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Benzothiazolyl)-3,5-diphenyltetrazolium Bromide 95.0+%, TCI America™
SDP

Catalog No. B0333100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0333100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0333100MG Supplier TCI America Supplier No. B0333100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 55699-64-4
Molecular Formula C20H14BrN5S
MDL Number MFCD00135111

Specifications

Quantity 100 mg
Formula Weight 436.33
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Benzothiazolyl)-3,5-diphenyltetrazolium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.