Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Bibenzothiazoline 85.0+%, TCI America™
SDP

Catalog No. B03771G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B03771G 1 g
B03775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B03771G Supplier TCI America Supplier No. B03771G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19258-20-9
Molecular Formula C14H12N2S2
Molecular Weight (g/mol) 272.384
MDL Number MFCD00059699
InChI Key LYPDJBMOMMJHAR-UHFFFAOYSA-N
PubChem CID 253953
IUPAC Name 2-(2,3-dihydro-1,3-benzothiazol-2-yl)-2,3-dihydro-1,3-benzothiazole
SMILES C1=CC=C2C(=C1)NC(S2)C3NC4=CC=CC=C4S3

Specifications

Melting Point 135°C
Quantity 1 g
Synonym 2,2′C-Bisbenzothiazoline, 2,2′C-Dibenzothiazoline
Formula Weight 272.38
Percent Purity ≥85.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bibenzothiazoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.