Learn More
Matrix Scientific 2,2'-Biimidazole, 492-98-8, MFCD00047014, 1g
Supplier: Matrix Scientific 0075131G
Molecular Formula C6H6N4, Purity 96%, Molecular Weight 134.14, Melting Point >350°
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.