Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Biphenylyl)amino-9,9-dimethylfluorene 98.0+%, TCI America™
SDP

Catalog No. B51221G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51221G 1 g
B51225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51221G Supplier TCI America Supplier No. B51221G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1198395-24-2
Molecular Formula C27H23N
Molecular Weight (g/mol) 361.49
MDL Number MFCD19440968
InChI Key OBARUOOPPWHZRQ-UHFFFAOYSA-N
Synonym N-([1,1′-Biphenyl]-2-yl)-9,9-dimethyl-9H-fluoren-2-amine
PubChem CID 66803057
IUPAC Name N-{[1,1'-biphenyl]-2-yl}-9,9-dimethyl-9H-fluoren-2-amine
SMILES CC1(C)C2=CC=CC=C2C2=C1C=C(NC1=CC=CC=C1C1=CC=CC=C1)C=C2

Specifications

Melting Point 120°C
Color White-Yellow
Quantity 1 g
Formula Weight 361.49
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Biphenylyl)amino-9,9-dimethylfluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.