Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2'-bipyridine-4-carboxylic acid

Catalog No. CDS018251 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS018251 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS018251 Supplier Sigma Aldrich Supplier No. CDS01825110MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • The product is a 70% solution of glycolic acid nitrile in water
  • Glycolic acid nitrile, also known as hydroxyacetonitrile is the simplest cyanohydrin
  • The microwave spectra of hydroxyacetonitrile have been recorded in gas-phase and its rotational constants have been calculated
  • Ice containing formaldehyde and hydrogen cyanide react under astrophysical-like conditions to form hydroxyacetonitrile

Specifications

Molecular Formula C11H8N2O2
Linear Formula C11H8N2O2
MDL Number MFCD09414714
Quantity 50 mL
Molecular Weight (g/mol) 200.19
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.