Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Bipyridine-6,6'-diol 98.0+%, TCI America™
SDP

Catalog No. B39661G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39661G 1 g
B39665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39661G Supplier TCI America Supplier No. B39661G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 103505-54-0
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.186
MDL Number MFCD13194839
InChI Key POJMWJLYXGXUNU-UHFFFAOYSA-N
PubChem CID 363601
IUPAC Name 6-(6-oxo-1H-pyridin-2-yl)-1H-pyridin-2-one
SMILES C1=CC(=O)NC(=C1)C2=CC=CC(=O)N2

Specifications

Melting Point 340°C
Color Yellow
Quantity 1 g
Synonym 6,6′C-Dihydroxy-2,2′C-bipyridine, 6,6′C-Dihydroxy-2,2′C-bipyridyl
Formula Weight 188.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bipyridine-6,6′-diol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.