Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Bipyridine-6-carbonitrile 98.0+%, TCI America™
SDP

Catalog No. B4557200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4557200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4557200MG Supplier TCI America Supplier No. B4557200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nitrogen-donor ligand used as a heterocyclic building block in the synthesis of functional metal complexes.

Specifications

CAS 4392-85-2
Melting Point 133°C
Molecular Formula C11H7N3
Quantity 200 mg
MDL Number MFCD07364147
Synonym 6-Cyano-2,2′C-bipyridyl, 6-Cyano-2,2′C-bipyridine
InChI Key QLZHLNAALBJVOE-UHFFFAOYSA-N
SMILES N#CC1=NC(=CC=C1)C1=CC=CC=N1
IUPAC Name [2,2'-bipyridine]-6-carbonitrile
Molecular Weight (g/mol) 181.20
PubChem CID 11095236
Formula Weight 181.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bipyridine-6-carbonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.