Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich [2,2'-Bipyridine]-6-Carboxylic Acid Hydrochloride

Catalog No. 901251500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-125-1500MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-125-1500MG Supplier Sigma Aldrich Supplier No. 901251500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Developed in the Engle lab, this 2,2'-bipyridylamide (PPA) is a pincer-like, removable tridentate directing group that stabilizes 6-membered palladacycles for olefin functionalization. Together with similar ligand PAQ (901250), researchers demonstrated regioselective remote hydrocarbofunctionalization of alkene-containing substrate classes (e.g. 4-pentenoic acids, allylic alcohols, homoallyl amines, bis-homoallylamines) using Pd(II) catalysis.Tridentate Directing Groups Stabilize 6-Membered Palladacycles in Catalytic Alkene Hydrofunctionalization

Specifications

CAS 2284459-08-9
Molecular Formula C32H20B4N6O3
Linear Formula C34H54ClN5O9S
Quantity 100 mg
Molecular Weight (g/mol) 579.78 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.