Learn More
Medchemexpress LLC 2,2'-bipyridine-d8 | 32190-42-4 | 99.3% | 164.23 g·mol⁻1 | C10D8N2 | 200 MG
SDP
Supplier:  Medchemexpress LLC HYW440409200MG
Encompass_Preferred
2,2'-Bipyridine-d8 is the deuterium-labeled analogue of 2,2'-bipyridine supplied as a high-purity solid for use as a stable-isotope reagent and analytical standard. It is intended for mechanistic studies, tracer experiments, and as an internal standard in mass-spectrometry workflows where deuterium labeling improves tracking and quantification.
  • High isotopic purity for accurate mass-spectrometry analysis.
  • Suitable as an internal standard and tracer in analytical workflows.
  • Solid form convenient for storage and handling.
  • Commonly used in coordination chemistry and ligand studies.
  • Supplied in a 200 mg pack size suitable for small-scale experiments.
Catalog No. 50-002-81170