Learn More
Medchemexpress LLC 2,2'-bipyridine-d8 | 32190-42-4 | 99.3% | 164.24 g/mol | C10D8N2 | 1 MG
SDP
Catalog No. 5000281146
Encompass_Preferred
2,2′-Bipyridine-d8 is the deuterium-labeled isotopologue of 2,2′-bipyridine designed for use as a tracer or internal standard in quantitative analytical techniques such as NMR, GC-MS, and LC-MS. Sold for research use only, it is supplied in small-quantity formats for analytical and method development applications.
  • Deuterium-labeled isotopologue suitable as tracer or internal standard.
  • High purity (99.3%).
  • Molecular formula C10D8N2; molecular weight 164.24 g/mol.
  • Available in small quantities (1 mg) for analytical use.
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-81146 Supplier Medchemexpress LLC Supplier No. HYW4404091MG
2,2′-Bipyridine-d8 is the deuterium-labeled isotopologue of 2,2′-bipyridine designed for use as a tracer or internal standard in quantitative analytical techniques such as NMR, GC-MS, and LC-MS. Sold for research use only, it is supplied in small-quantity formats for analytical and method development applications.
  • Deuterium-labeled isotopologue suitable as tracer or internal standard.
  • High purity (99.3%).
  • Molecular formula C10D8N2; molecular weight 164.24 g/mol.
  • Available in small quantities (1 mg) for analytical use.
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.