Learn More
Medchemexpress LLC 2,2′-bipyridine-d8 | 32190-42-4 | MFCD00190432 | 99.3% | 164.24 g·mol⁻¹ | C10D8N2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW44040950MG
Encompass_Preferred
2,2′-Bipyridine-d8 is the deuterium-labeled isotopologue of 2,2′-bipyridine used as a stable isotope tracer and analytical standard in chemical and biochemical research. It is supplied as a solid for laboratory use and is intended for research applications including mass spectrometry, mechanistic studies, and ligand chemistry.
  • Deuterium labeled for use as a tracer and internal standard.
  • High isotopic enrichment and chemical purity suitable for analytical work.
  • Useful in mass spectrometry and NMR experiments to simplify spectral interpretation.
  • Solid form, easy to weigh and dissolve in common organic solvents.
  • Supplied with certificate of analysis and safety data sheet for quality assurance.
Catalog No. 50-002-81471