Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2'-Bipyridyl

Catalog No. D21630525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21630525G 25 g
D216305100G 100 g
D21630510G 10 g
D2163051KG 1 kg
D216305500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. D21630525G Supplier Sigma Aldrich Supplier No. D21630525G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,2'-Bipyridyl

Specifications

CAS 366-18-7
Melting Point 70°C to 73°C
Boiling Point 273°C
Molecular Formula C10H8N2
Linear Formula C10H8N2
MDL Number MFCD00006212
Quantity 25 g
Synonym 2,2'-Bipyridine; 2,2'-Dipyridine; 2,2'-Dipyridyl
Molecular Weight (g/mol) 156.18 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.