Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Bipyridyl-d8 98atom%D 99.0+%, TCI America™
SDP

Catalog No. B5258200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5258200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B5258200MG Supplier TCI America Supplier No. B5258200MG
Add to Cart
Add to Cart

Deuterated reagent with 98 atom% D enrichment for analytical and research applications.

Chemical Identifiers

CAS 32190-42-4
Molecular Formula C10H8N2
Molecular Weight (g/mol) 164.237
MDL Number MFCD00190432
InChI Key ROFVEXUMMXZLPA-PGRXLJNUSA-N
Synonym 2,2′-Bipyridine-d8, Octadeutero-2,2′-bipyridyl
PubChem CID 11378683
IUPAC Name 2,3,4,5-tetradeuterio-6-(3,4,5,6-tetradeuteriopyridin-2-yl)pyridine
SMILES C1=CC=NC(=C1)C2=CC=CC=N2

Specifications

Melting Point 72°C
Boiling Point 273°C
Quantity 200 mg
UN Number 2811
Formula Weight 164.24
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bipyridyl-d8 98atom%D
EINECSNumber (5)-3723
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.