Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Bis(2-oxazoline) 97.0+%, TCI America™
SDP

Catalog No. B12265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12265G 5 g
B122625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B12265G Supplier TCI America Supplier No. B12265G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 36697-72-0
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.14
MDL Number MFCD00059930
InChI Key KKKKCPPTESQGQH-UHFFFAOYSA-N
PubChem CID 283385
IUPAC Name 2-(4,5-dihydro-1,3-oxazol-2-yl)-4,5-dihydro-1,3-oxazole
SMILES C1CN=C(O1)C1=NCCO1

Specifications

Melting Point 213°C
Quantity 5 g
Formula Weight 140.14
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bis(2-oxazoline)
EINECSNumber (5)-5584
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.